3-Cyano-4-methylpyridine ≥98.0%
Leverantör: TCI
Varning
Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
C2608-5G C2608-1G
TCIAC2608-5GEA 1725 SEK
TCIAC2608-5G TCIAC2608-1G
3-Cyano-4-methylpyridine ≥98.0%
3-Cyano-4-methylpyridine
Formel: C₇H₆N₂
Kokpunkt: 111°C
MDL Number: MFCD00234272
CAS nummer: 5444-01-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f5b6bbedf3df51d2080e7511957ef69e6345688a&vid=f5b6bbedf3df51d2080e7511957ef69e6345688a&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 45.0 to 49.0 deg-C