(-)-Quinine hydrochloride dihydrate 99% may contain up to 10% Dihydroquinine
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: Quinine hydrochloride dihydrate, Quinine monohydrochloride dihydrate, (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride dihydrate
H33474.09 H33474.18
H33474.09EA 840 SEK
H33474.09 H33474.18
(-)-Quinine hydrochloride dihydrate 99% may contain up to 10% Dihydroquinine
(-)-Quinine hydrochloride dihydrate
Purity: 99% CAS No.: 6119-47-7 Molecular Formula: C20H24N2O2.HCl.2H2O Formula Weight: 396.91 (360.88anhy) Beilstein Reference: 6112655 Merck Reference: 14,8061 Optical Rotation: -246° (c=2 in 0.1M HCl) MDL No.: MFCD00151248
Formel: C₂₀H₂₅ClN₂O₂·2H₂O
Smältpunkt: 158…160 °C
Flampunkt: 122 °C
Storage Temperature: Omgivande
MDL Number: MFCD00151248
CAS nummer: 6119-47-7
EINECS: 205-001-1
Merck Index: 14,8061