4-Methoxystyrene ≥98.0% stabilised

Leverantör: TCI EUROPE
Varning

Synonyms: 1-Methoxy-4-vinylbenzene, 4-Vinylanisole

Stabilization: med 4-tert-Butylpyrokatekol

M0130-10ML M0130-25ML
TCIAM0130-10MLEA 1150 SEK
TCIAM0130-10ML TCIAM0130-25ML
4-Methoxystyrene ≥98.0% stabilised
4-Methoxystyrene
Formel: C₉H₁₀O
MW: 134,18 g/mol
Kokpunkt: 98°C
Smältpunkt: 3,3 °C
Densitet: 1,002 g/cm³ (20 °C)
Flampunkt: 76°C
Storage Temperature: Kylskåp
MDL Number: MFCD00008619
CAS nummer: 637-69-4
EINECS: 211-298-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=099c113b17369dd7fc467c6b3a982e828b1871ea&vid=099c113b17369dd7fc467c6b3a982e828b1871ea&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR