Bortriklorid 1 M i diklormetan stabilised
Leverantör: Thermo Fisher Scientific
Fara
H32751.AE H32751.AP
H32751.AEEA 2100 SEK
H32751.AE H32751.AP
Bortriklorid 1 M i diklormetan stabilised
Bortriklorid
CAS No.: 10294-34-5 Molecular Formula: BCl3 Formula Weight: 117.17 Storage Temperature: 2°C to 8°C UN No.: 2927 MDL No.: MFCD00011313
Formel: BCl₃
MW: 117.17 g/mol
Kokpunkt: 12.4
Smältpunkt: -107
Densitet: 1.326
Storage Temperature: Kylskåp
MDL Number: MFCD00011313
CAS nummer: 10294-34-5
EINECS: 233-658-4
UN: 2927
ADR: 6.1 II
Merck Index: 12,01378