Cyclopentyl isocyanate ≥98.0%
Leverantör: TCI
Fara
C2543-5G C2543-1G
TCIAC2543-5GEA 1050 SEK
TCIAC2543-5G TCIAC2543-1G
Cyclopentyl isocyanate ≥98.0%
Cyclopentyl isocyanate
Formel: C₆H₉NO
MW: 111.14 g/mol
Kokpunkt: 146°C
Flampunkt: 36°C
Storage Temperature: Kylskåp
MDL Number: MFCD02093695
CAS nummer: 4747-71-1
UN: 3080
ADR: 6.1 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2284cb4d6277b05895b968a42b1ea3964310f5e5&vid=2284cb4d6277b05895b968a42b1ea3964310f5e5&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %