Dimethyl-5-bromoisophthalate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Dimethyl 5-bromoisophthalate, 5-Bromoisophthalic acid dimethyl ester

D4068-1G D4068-5G
TCIAD4068-1GEA 335 SEK
TCIAD4068-1G TCIAD4068-5G
Dimethyl-5-bromoisophthalate ≥98.0%
Dimethyl-5-bromoisophthalate
Formel: C₁₀H₉BrO₄
MW: 273.08 g/mol
Kokpunkt: 159°C
MDL Number: MFCD00078709
CAS nummer: 51760-21-5
EINECS: 257-386-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e7047e22edba37653532fd7a23e242303c391d0b&vid=e7047e22edba37653532fd7a23e242303c391d0b&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR