Guanidinhydroklorid
Leverantör: G Biosciences
Varning
Synonyms: Guanadine hydrochloride, Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidine HCI, Guanidine chloride, Guanidinium chloride, Guanidine hydrochloride
BC88 BC85 RC-057
BC88EA 5250 SEK
BC88 BC85 RC-057
Guanidinhydroklorid
Guanidinhydroklorid
A selection of strong chemicals to aid in the denaturation of proteins. Denaturants, or chaotropes, disrupt water interactions resulting in the solubilization of hydrophobic proteins and peptides. The chaotropes also act on all proteins to unfold them and alter their three-dimensional structure.
Available as a powder or 8M solution
Formel: NH₂C(=NH)NH₂·HCl
MW: 95.53 g/mol
Smältpunkt: 185
Densitet: 1.354 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00013026
CAS nummer: 50-01-1
EINECS: 200-002-3
Merck Index: 13,04578
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da5a871cab977e6d9dc666b457732eb1e869a714&vid=b0d4fcb044b6106fce8e40448dbf2ce6e7077ad7&prid=web&clid=SAPDAM