Indol-3-acetic syra ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: IAA, 1H-Indole-3-acetic acid, Heteroauxin, 3-Indoleacetic acid
I0022-25G I0022-5G
TCIAI0022-25GEA 590 SEK
TCIAI0022-25G TCIAI0022-5G
Indol-3-acetic syra ≥98.0%
Indol-3-acetic syra
Formel: C₁₀H₉NO₂
MW: 175,19 g/mol
Kokpunkt: 120 - 125 °C (0,05 -rr)
Smältpunkt: 167…170 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00005636
CAS nummer: 87-51-4
EINECS: 201-748-2
Merck Index: 14,04964
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=890c3756018884834d6ef23cd68d83fd25db4919&vid=09dbc7b35ae84423dd34698978cbb25789b98c61&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Yellow to Orange powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 166.0 to 170.0 deg-C
Solubility in Methanol almost transparency