Strontium isopropoxide ≥96%
Leverantör: Thermo Scientific
Fara
Synonyms: Strontium i-propoxide
14481.03 14481.06 14481.14
14481.03EA 1600 SEK
14481.03 14481.06 14481.14
Strontium isopropoxide ≥96%
Strontium isopropoxide
Purity: 96% CAS No.: 88863-33-6 Molecular Formula: Sr[OCH(CH3)2]2 Formula Weight: 205.82 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: 3205 MDL No.: MFCD00050487
Formel: C₆H₁₄O₂Sr
MW: 205,79 g/mol
Smältpunkt: 130 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00050487
CAS nummer: 88863-33-6
UN: 3205
ADR: 4.2,III