Barium isopropoxide 20% (w/v) i 2-propanol
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Barium i-propoxide
41338.AB 41338.AD
41338.ABEA 1900 SEK
41338.AB 41338.AD
Barium isopropoxide 20% (w/v) i 2-propanol
Barium isopropoxide
CAS No.: 24363-37-9 Molecular Formula: C6H14BaO2 Formula Weight: 255.52 Flash Point: 22°(72°F) Physical Form: Liquid Density: 0.89 Appearance: Yellow Odour: Alcohol-like UN No.: 3274 MDL No.: MFCD00050486
In Sol-Gel synthesis and superconductors
Formel: C₆H₁₄BaO₂
MW: 255.501 g/mol
Densitet: 0.89
Flampunkt: 22 °C (72 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00050486
CAS nummer: 24363-37-9
UN: 3274
ADR: 3 II