3,3'-Dimethyldiphenylamine ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Di-m-tolylamine

D1512-25G D1512-5G
TCIAD1512-25GEA 3925 SEK
TCIAD1512-25G TCIAD1512-5G
3,3'-Dimethyldiphenylamine ≥98.0%
3,3'-Dimethyldiphenylamine
Formel: C₁₄H₁₅N
MW: 197,28 g/mol
Kokpunkt: 200°C
Smältpunkt: –12°C
MDL Number: MFCD00059315
CAS nummer: 626-13-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e6e85f0a40a92e812d4c047bc75b52752be5882e&vid=e6e85f0a40a92e812d4c047bc75b52752be5882e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR