α,α,α,2,4-Pentachlorotoluene ≥99.0%
Leverantör: TCI
Varning
Synonyms: 2,4-Dichlorobenzotrichloride
D0428-25G D0428-500G
TCIAD0428-25GEA 215 SEK
TCIAD0428-25G TCIAD0428-500G
α,α,α,2,4-Pentachlorotoluene ≥99.0%
α,α,α,2,4-Pentachlorotoluene
Formel: C₇H₃Cl₅
Kokpunkt: 159°C
MDL Number: MFCD00018818
CAS nummer: 13014-18-1
EINECS: 235-868-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=866de951276a75ae16cb577cf766cb705f388fe0&vid=866de951276a75ae16cb577cf766cb705f388fe0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 46.0 to 50.0 deg-C