4-Phenoxyphenylboronic acid 95+%
Leverantör: Thermo Scientific
Varning
Synonyms: (4-Phenoxyphenyl)boronic acid, 4-Phenoxybenzeneboronic acid
359640010 359640050 359640250
ACRO359640010EA 680 SEK
ACRO359640010 ACRO359640050 ACRO359640250
4-Phenoxyphenylboronic acid 95+%
4-Phenoxyphenylboronic acid
Formel: C₁₂H₁₁BO₃
MW: 214,03 g/mol
Smältpunkt: 141…145 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00093312
CAS nummer: 51067-38-0
Order Now
Specifikation testresultat
Appearance White to off-white Powder and/or chunks
Infrared spectrum Conforms
HPLC ≥95.0 %