4-Chloro-6-nitro-o-toluidine ≥98%
Leverantör: Thermo Scientific
Fara
Synonyms: 4-Chloro-2-methyl-6-nitroaniline
B22253.03 B22253.06
B22253.03EA 765 SEK
B22253.03 B22253.06
4-Chloro-6-nitro-o-toluidine ≥98%
4-Chloro-6-nitro-o-toluidine
Purity: 98% CAS No.: 62790-50-5 Molecular Formula: C7H7ClN2O2 Formula Weight: 186.60 Melting Point: 121-123° MDL No.: MFCD01320687
Formel: C₇H₇ClN₂O₂
MW: 186,60 g/mol
Smältpunkt: 121…123 °C
Storage Temperature: Omgivande
MDL Number: MFCD01320687
CAS nummer: 62790-50-5
UN: 2237
ADR: 6.1,III