2-Chloroethyl chloroformate ≥98.0%
Leverantör: TCI EUROPE
Fara
Synonyms: 2-Chloroethyl carbonochloridate
C1052-25G C1052-100G
TCIAC1052-25GEA 390 SEK
TCIAC1052-25G TCIAC1052-100G
2-Chloroethyl chloroformate ≥98.0%
2-Chloroethyl chloroformate
Formel: C₃H₄Cl₂O₂
MW: 142,9687 g/mol
Kokpunkt: 154°C
Flampunkt: 60°C
Storage Temperature: Kylskåp
MDL Number: MFCD00000646
CAS nummer: 627-11-2
EINECS: 210-982-4
UN: 2742
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d11753bc1e2cf00738c390cf943f7bcc3bdd58f6&vid=d11753bc1e2cf00738c390cf943f7bcc3bdd58f6&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %
Purity (Argentmetric Titration) min. 98.0 %