Trimethylamine-N-oxide dihydrate ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Trimethylamine oxide dihydrate

T0466-25G T0466-500G
TCIAT0466-25GEA 390 SEK
TCIAT0466-25G TCIAT0466-500G
Trimethylamine-N-oxide dihydrate ≥98.0%
Trimethylamine-N-oxide dihydrate
Formel: C₃H₉NO·2H₂O
MW: 111,14 g/mol (75,11 anhy)
MDL Number: MFCD00002048
CAS nummer: 62637-93-8
EINECS: 214-675-6
Merck Index: 14,09711
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=709b62d620052f88591e91ac90efe6bc1c46ce28&vid=709b62d620052f88591e91ac90efe6bc1c46ce28&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 95.0 to 99.0 deg-C
Solubility in Water almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR