Methyl pentafluorobenzoate ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: Methyl Pentafluorobenzoate
P1418-25G P1418-5G
TCIAP1418-25GEA 1350 SEK
TCIAP1418-25G TCIAP1418-5G
Methyl pentafluorobenzoate ≥98.0%
Methyl pentafluorobenzoate
Formel: C₈H₃F₅O₂
MW: 226,1002 g/mol
Flampunkt: 78°C
MDL Number: MFCD00012172
CAS nummer: 36629-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f253072b4b80bc30af38bbeee265d614d29a12df&vid=f253072b4b80bc30af38bbeee265d614d29a12df&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %