Ethyl pentafluoropropionate ≥98.0%

Leverantör: TCI EUROPE
Fara

P1123-25G P1123-5G
TCIAP1123-25GEA 1150 SEK
TCIAP1123-25G TCIAP1123-5G
Ethyl pentafluoropropionate ≥98.0%
Ethyl pentafluoropropionate
Formel: C₅H₅F₅O₂
MW: 192,09 g/mol
Kokpunkt: 76°C
Flampunkt: 2°C
MDL Number: MFCD00000431
CAS nummer: 426-65-3
EINECS: 207-043-6
UN: 3272
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae81744a5b5bbc4e7346d4855c56d6eb3d1cdd15&vid=ae81744a5b5bbc4e7346d4855c56d6eb3d1cdd15&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR