Ethyl chlorofluoroacetate ≥98.0%

Leverantör: TCI
Fara

Synonyms: Ethylchlorofluoroacetate

C1467-25G C1467-5G
TCIAC1467-25GEA 1475 SEK
TCIAC1467-25G TCIAC1467-5G
Ethyl chlorofluoroacetate ≥98.0%
Ethyl chlorofluoroacetate
Formel: C₄H₆ClFO₂
MW: 140.54 g/mol
Kokpunkt: 130°C
Flampunkt: 23°C
MDL Number: MFCD00039333
CAS nummer: 401-56-9
EINECS: 206-930-5
UN: 2924
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bc65dc508c10845035a7f3bd98c4d85550c050d&vid=2bc65dc508c10845035a7f3bd98c4d85550c050d&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR