Benzyl-4-bromophenyl ketone ≥98%
Leverantör: Thermo Scientific
Synonyms: 1-(4-Bromophenyl)-2-phenylethanone
B22159.03 B22159.06
B22159.03EA 1075 SEK
B22159.03 B22159.06
Benzyl-4-bromophenyl ketone ≥98%
Benzyl-4-bromophenyl ketone
Purity: 98% CAS No.: 2001-29-8 Molecular Formula: C14H11BrO Formula Weight: 275.14 Melting Point: 109-112° Beilstein Reference: 2211322
Formel: C₁₄H₁₁BrO
MW: 275,14 g/mol
Smältpunkt: 109…112 °C
Storage Temperature: Omgivande
MDL Number: MFCD00016331
CAS nummer: 2001-29-8