4-(Trifluormethyl)nicotinonitrile

Leverantör: Thermo Scientific

RF04123DA RF04123EA
MAYBRF04123DAEA 0 SEK
MAYBRF04123DA MAYBRF04123EA
4-(Trifluormethyl)nicotinonitrile
4-(Trifluormethyl)nicotinonitrile
Formel: C₇H₃F₃N₂
MDL Number: MFCD00153107
CAS nummer: 13600-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR