6-Aminonicotinamide ≥99.0%
Leverantör: TCI
|
Fara
|
Synonyms:
6-Aminopyridine-3-carboxamide
A0986-1G
A0986-5G
TCIAA0986-1GEA
950
SEK
TCIAA0986-1G
TCIAA0986-5G
6-Aminonicotinamide ≥99.0%
6-Aminonicotinamide
Formel:
C₆H₇N₃O MW: 137.14 g/mol |
MDL Number:
MFCD00006327 CAS nummer: 329-89-5 EINECS: 206-349-7 UN: 2811 ADR: 6.1 III |
|
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (Nonaqueous Titration) | min. 99.0 % |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...