6-Chloronicotinoyl chloride ≥98.0%

Leverantör: TCI
Fara

C1726-25G C1726-5G
TCIAC1726-25GEA 1150 SEK
TCIAC1726-25G TCIAC1726-5G
6-Chloronicotinoyl chloride ≥98.0%
6-Chloronicotinoyl chloride
Formel: C₆H₃Cl₂NO
MDL Number: MFCD00051775
CAS nummer: 58757-38-3
UN: 3261
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86d8530f757a2bbfb6a379f52393c740c556fbb6&vid=86d8530f757a2bbfb6a379f52393c740c556fbb6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 47.0 and le 51.0 deg-C
Solubility in Toluene very faint turbidity


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR