4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%
Leverantör: Thermo Fisher Scientific
Varning
L16150.06 L16150.14 L16150.22
L16150.06EA 1625 SEK
L16150.06 L16150.14 L16150.22
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride ≥99%
4,4'-(Hexafluoroisopropylidene)diphthalic anhydride
Purity: 99% CAS No.: 1107-00-2 Molecular Formula: C19H6F6O6 Formula Weight: 444.25 Melting Point: 244-247° Beilstein Reference: 7057916
Formel: C₈H₄O₃
Smältpunkt: 244…247 °C
Storage Temperature: Omgivande
MDL Number: MFCD00039143
CAS nummer: 1107-00-2
EINECS: 214-170-0