2-Morpholino-3-pyridinamine

Leverantör: Thermo Scientific

Synonyms: 3-Amino-2-morpholin-4-ylpyridine

CC24614DA CC24614EA
MAYBCC24614DAEA 0 SEK
MAYBCC24614DA MAYBCC24614EA
2-Morpholino-3-pyridinamine
2-Morpholino-3-pyridinamine
Formel: C₉H₁₃N₃O
MDL Number: MFCD03001243
CAS nummer: 51627-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR