2',3'-Dihydroxy-4'-methoxyacetophenone hydrate ≥98.0%
Leverantör: TCI
|
Varning
|
D2187-25G
D2187-5G
TCIAD2187-25GEA
2525
SEK
TCIAD2187-25G
TCIAD2187-5G
2',3'-Dihydroxy-4'-methoxyacetophenone hydrate ≥98.0%
2',3'-Dihydroxy-4'-methoxyacetophenone hydrate
Formel:
C₉H₁₀O₄·H₂O |
MDL Number:
MFCD00180772 CAS nummer: 708-53-2 |
|
Specifikation testresultat
Appearance | Light yellow to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 95.0 % |
Melting point | 130.0 to 135.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...