Methyl-4-acetylbenzoate ≥98.0%

Leverantör: TCI

Synonyms: Methyl 4-acetylbenzoate

M2163-25G M2163-5G
TCIAM2163-25GEA 2850 SEK
TCIAM2163-25G TCIAM2163-5G
Methyl-4-acetylbenzoate ≥98.0%
Methyl-4-acetylbenzoate
Formel: C₁₀H₁₀O₃
MDL Number: MFCD00216474
CAS nummer: 3609-53-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd15b3d1f9977f9263de1a59912f6f4a0eec66cf&vid=dd15b3d1f9977f9263de1a59912f6f4a0eec66cf&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 93.0 to 97.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR