Piceatannol 98,0 HPLC_ANALYSMETOD

Leverantör: TCI

Synonyms: (E)-4-(3,5-Dihydroxystyryl)benzene-1,2-diol

P1928-1G P1928-100MG
TCIAP1928-1GEA 5825 SEK
TCIAP1928-1G TCIAP1928-100MG
Piceatannol 98,0 HPLC_ANALYSMETOD
Piceatannol
Formel: C₁₄H₁₂O₄
MW: 244.2427 g/mol
Kokpunkt: 108°C
Smältpunkt: 235°C
Storage Temperature: Kylskåp
MDL Number: MFCD00221715
CAS nummer: 10083-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=85d032f03f8ce5cd6a2f49cd44a2927f07f3224e&vid=85d032f03f8ce5cd6a2f49cd44a2927f07f3224e&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light red powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR