3-Methoxydiphenylamine ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Methoxy-N-phenylaniline
M1135-25G
M1135-500G
TCIAM1135-25GEA
310
SEK
TCIAM1135-25G
TCIAM1135-500G
3-Methoxydiphenylamine ≥98.0%
3-Methoxydiphenylamine
Formel:
C₁₃H₁₃NO MW: 199,25 g/mol Kokpunkt: 160°C |
MDL Number:
MFCD00008383 CAS nummer: 101-16-6 EINECS: 202-921-5 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 71.0 to 74.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...