4-(Phenylazo)diphenylamine 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: 4-Phenylazodiphenylamine
212010050 212010250
ACRO212010050EA 535 SEK
ACRO212010050 ACRO212010250
4-(Phenylazo)diphenylamine 97%
4-(Phenylazo)diphenylamine
Formel: C₁₈H₁₅N₃
MW: 273.34 g/mol
Smältpunkt: 84…91 °C
Storage Temperature: Omgivande
MDL Number: MFCD00003023
CAS nummer: 101-75-7
EINECS: 202-972-3
Order Now
Specifikation testresultat
Appearance Orange to brown Powder
Infrared spectrum Conforms
Melting point 89 to 91 °C
HPLC ≥96.0 %