1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97.0% hydroxyl protecting agent

Leverantör: TCI EUROPE
Fara

Synonyms: 1,3-Dichloro-1,1,3,3-tetrakis(isopropyl)disiloxane

D1608-25G D1608-5G
TCIAD1608-25GEA 2600 SEK
TCIAD1608-25G TCIAD1608-5G
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane ≥97.0% hydroxyl protecting agent
1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane
Formel: C₁₂H₂₈Cl₂OSi₂
MW: 315,43 g/mol
Kokpunkt: 90°C
Flampunkt: 76°C
MDL Number: MFCD00009655
CAS nummer: 69304-37-6
UN: 2987
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e9ef74f77f03e5987bdc32a18b9ad245b9713950&vid=82a558ac2f5610e031870bd6a5557e4b4915cb60&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR