1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane ≥97%
Leverantör: Thermo Scientific
Fara
L16605.06 L16605.14
L16605.06EA 1450 SEK
L16605.06 L16605.14
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane ≥97%
1H,1H,2H,2H-Perfluorooctylmethyldichlorosilane
Purity: 97% CAS No.: 73609-36-6 Molecular Formula: C9H7Cl2F13Si Formula Weight: 461.13 Flash Point: 51°(123°F) Refractive Index: 1.3500 Boiling Point: 189-190° Beilstein Reference: 6153667 UN No.: 2920 MDL No.: MFCD00042360
Formel: C₉H₇Cl₂F₁₃Si
MW: 461,13 g/mol
Kokpunkt: 189…190 °C
Densitet: 1,550
Flampunkt: 51 °C (123 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00042360
CAS nummer: 73609-36-6
EINECS: 277-551-0
UN: 2920
ADR: 8,II