1,1,2-Trimethyl-1H-benz[e]indole ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 1,1,2-Trimethyl-1H-benzo[e]indole

T1714-25G T1714-5G T1714-100G
TCIAT1714-25GEA 1350 SEK
TCIAT1714-25G TCIAT1714-5G TCIAT1714-100G
1,1,2-Trimethyl-1H-benz[e]indole ≥98.0% (by HPLC, titration analysis)
1,1,2-Trimethyl-1H-benz[e]indole
Formel: C₁₅H₁₅N
MW: 209,29 g/mol
MDL Number: MFCD00082627
CAS nummer: 41532-84-7
EINECS: 255-429-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4b2f2ad2d9c4841c7528b870897d423f060fe633&vid=4b2f2ad2d9c4841c7528b870897d423f060fe633&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 115.0 to 119.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR