1,1,2-Trimethyl-1H-benz[e]indole ≥97%
Leverantör: Thermo Scientific
|
Varning
|
Synonyms:
1,1,2-Trimethyl-1H-benzo[e]indole
B21804.06
B21804.14
B21804.06EA
945
SEK
B21804.06
B21804.14
1,1,2-Trimethyl-1H-benz[e]indole ≥97%
1,1,2-Trimethyl-1H-benz[e]indole
Purity: 97%
CAS No.: 41532-84-7
Molecular Formula: C15H15N
Formula Weight: 209.29
Melting Point: 114-119°
Beilstein Reference: 153709
Formel:
C₁₅H₁₅N MW: 209,29 g/mol Smältpunkt: 114…119 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00082627 CAS nummer: 41532-84-7 EINECS: 255-429-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...