Dimethylphenacylsulphonium tetrafluoroborate ≥98.0%

Leverantör: TCI
Fara

D2231-1G D2231-5G
TCIAD2231-1GEA 1150 SEK
TCIAD2231-1G TCIAD2231-5G
Dimethylphenacylsulphonium tetrafluoroborate ≥98.0%
Dimethylphenacylsulphonium tetrafluoroborate
Formel: C₁₀H₁₃BF₄OS
MDL Number: MFCD00191352
CAS nummer: 24806-57-3
UN: 1759
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e62724214cacb5e655d0bb0e5b7714dbbb710c46&vid=e62724214cacb5e655d0bb0e5b7714dbbb710c46&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 169.0 to 172.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR