(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide
D2164-25G
D2164-5G
TCIAD2164-25GEA
1550
SEK
TCIAD2164-25G
TCIAD2164-5G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide ≥97.0%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide
Formel:
C₂₂H₂₂BrO₂P MW: 429,30 g/mol |
MDL Number:
MFCD00011966 CAS nummer: 52509-14-5 EINECS: 257-977-3 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (Nonaqueous Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...