Cyclohexyl(dimethoxy)methylsilane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Cyclohexyldimethoxy(methyl)silane

C1982-25ML C1982-500ML
TCIAC1982-25MLEA 215 SEK
TCIAC1982-25ML TCIAC1982-500ML
Cyclohexyl(dimethoxy)methylsilane ≥98.0%
Cyclohexyl(dimethoxy)methylsilane
Formel: C₉H₂₀O₂Si
MW: 188,3394 g/mol
Kokpunkt: 196°C
Flampunkt: 66°C
MDL Number: MFCD00071752
CAS nummer: 17865-32-6
EINECS: 402-140-1
UN: 3082
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=20b21a34114426ebb7363aadfe4e5a93bde9e5ae&vid=20b21a34114426ebb7363aadfe4e5a93bde9e5ae&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR