Aluminium magnesium isopropoxide ≥99.95%
Leverantör: Thermo Fisher Scientific
Fara
Synonyms: Magnesium aluminum i-propoxide
39320.04 39320.09 39320.18
39320.04EA 1175 SEK
39320.04 39320.09 39320.18
Aluminium magnesium isopropoxide ≥99.95%
Aluminium magnesium isopropoxide
Purity: 99.95+% CAS No.: 69207-83-6 Molecular Formula: Mg[Al(OCH(CH3)2)4]2 Formula Weight: 550.53 Physical Form: Low melt crystalline mass Sensitivity: Moisture Sensitive Boiling Point: 114°/0.2mm UN No.: 3181 MDL No.: MFCD00070426
Formel: C₂₄H₅₆Al₂MgO₈
MW: 550.965 g/mol
Kokpunkt: 114 °C (0.2 mmHg)
Smältpunkt: 40 °C
Storage Temperature: Omgivande
MDL Number: MFCD00070426
CAS nummer: 69207-83-6
UN: 3181
ADR: 4.1 III