1,3,5-Triethynylbenzene ≥98%
Leverantör: Thermo Scientific
Varning
B22417.03 B22417.06
B22417.03EA 3550 SEK
B22417.03 B22417.06
1,3,5-Triethynylbenzene ≥98%
1,3,5-Triethynylbenzene
Purity: 98% CAS No.: 7567-63-7 Molecular Formula: C12H6 Formula Weight: 150.18 Melting Point: 105-107° UN No.: 1325 MDL No.: MFCD00068621
Formel: C₁₂H₆
MW: 150,18 g/mol
Smältpunkt: 105…107 °C
Densitet: 1,05
Storage Temperature: Omgivande
MDL Number: MFCD00068621
CAS nummer: 7567-63-7
UN: 1325
ADR: 4.1,III