Hydrochlorothiazide 97,0 HPLC, N
Leverantör: TCI
|
Varning
|
H1274-25G
H1274-5G
TCIAH1274-25GEA
1050
SEK
TCIAH1274-25G
TCIAH1274-5G
Hydrochlorothiazide 97,0 HPLC, N
Hydrochlorothiazide
Formel:
C₇H₈ClN₃O₄S₂ |
MDL Number:
MFCD00051765 CAS nummer: 58-93-5 EINECS: 200-403-3 Merck Index: 14,04781 |
|
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (with Total Nitrogen) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...