Retinoic acid 97%
Leverantör: Thermo Fisher Scientific
Varning
Synonyms: all-trans-Retinoic acid, (2E,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid, ATRA, Vitamin A acid, Tretinoin
207341000 207340010 207340050
ACRO207341000EA 785 SEK
ACRO207341000 ACRO207340010 ACRO207340050
Retinoic acid 97%
Retinoic acid
Formel: C₂₀H₂₈O₂
MW: 300.44 g/mol
Smältpunkt: 179…184 °C
Storage Temperature: Frys
MDL Number: MFCD00001551
CAS nummer: 302-79-4
EINECS: 206-129-0
Order Now
Specifikation testresultat
Appearance Yellow Crystalline powder
Infrared spectrum Conforms
Melting point 179 to 184 °C
HPLC ≥96.0 %