Copper(I) bromide dimethyl sulphide complex
Leverantör: Thermo Scientific
Varning
19368.04 19368.09 19368.18
19368.04EA 765 SEK
19368.04 19368.09 19368.18
Copper(I) bromide dimethyl sulphide complex
Copper(I) bromide dimethyl sulphide complex
CAS No.: 54678-23-8 Molecular Formula: CuBr.S(CH3)2 Formula Weight: 205.58 Physical Form: Crystalline Sensitivity: Air and Moisture Sensitive Appearance: Pale brown or pale green Melting Point: 124-129° dec. UN No.: 3335 MDL No.: MFCD00043295
Formel: C₂H₆BrCuS
MW: 205,58 g/mol
Smältpunkt: 124…129 °C (dec.)
Storage Temperature: Omgivande
MDL Number: MFCD00043295
CAS nummer: 54678-23-8
EINECS: 259-292-5
ADR: 9,III