(Methylthio)acetaldehyde dimethyl acetal ≥98.0%

Leverantör: TCI
Fara

Synonyms: (Methylthio)acetaldehydedimethylacetal

M2236-25G M2236-5G
TCIAM2236-25GEA 2475 SEK
TCIAM2236-25G TCIAM2236-5G
(Methylthio)acetaldehyde dimethyl acetal ≥98.0%
(Methylthio)acetaldehyde dimethyl acetal
Formel: C₅H₁₂O₂S
MW: 136.21 g/mol
Kokpunkt: 60°C
Flampunkt: –19°C
MDL Number: MFCD00009848
CAS nummer: 40015-15-4
UN: 1993
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0eacfa5d9e42634f13b0751ca66267b982f5ae6&vid=a0eacfa5d9e42634f13b0751ca66267b982f5ae6&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR