Calcium pyruvate, vattenfri 97%
Leverantör: Thermo Fisher Scientific
Synonyms: Calcium 2-oxopropanoate
B25204.14 B25204.22
B25204.14EA 615 SEK
B25204.14 B25204.22
Calcium pyruvate, vattenfri 97%
Calcium pyruvate
Purity: 97% CAS No.: 52009-14-0 Molecular Formula: C6H6CaO6 Formula Weight: 214.19 MDL No.: MFCD00037199
Formel: C₆H₆CaO₆
MW: 214.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00037199
CAS nummer: 52009-14-0
EINECS: 257-599-9