N-(3-Acetyl-4-hydroxyphenyl)butyramide ≥97%
Leverantör: Thermo Scientific
Synonyms: Ac-HoPhe-OH
A18672.18 A18672.0B
A18672.18EA 665 SEK
A18672.18 A18672.0B
N-(3-Acetyl-4-hydroxyphenyl)butyramide ≥97%
N-(3-Acetyl-4-hydroxyphenyl)butyramide
Purity: 97% CAS No.: 40188-45-2 Molecular Formula: C12H15NO3 Formula Weight: 221.26 Melting Point: 124-126° MDL No.: MFCD00798556
Formel: C₁₂H₁₅NO₃
MW: 221,26 g/mol
Smältpunkt: 124…126 °C
Storage Temperature: Omgivande
MDL Number: MFCD00798556
CAS nummer: 40188-45-2
EINECS: 254-831-0