Eshmuno® CPX, resin for racemat separation, för kromatografi
Leverantör: MERCK PRODUCTION CHEMICALS
|
Varning
|
1.20083.0100
1.20083.0500
1.20083.0100_T
1.20083.0500_T
1.20083.5000
1.20083.0100EA
34525
SEK
1.20083.0100
1.20083.0500
1.20083.0100_T
1.20083.0500_T
1.20083.5000
Eshmuno® CPX, resin for racemat separation, för kromatografi
Eshmuno® CPX
Eshmuno® CPX media is a strong cation exchanger built on the proven Eshmuno® resin technology.
- Excellent mAb monomer/aggregate separation efficiency
- High resolution intermediate purification
- Superior dynamic binding capacity
- High throughput purification
- Rigid base bead for easy packing
- Outstanding pressure flow behavior
Storage Temperature:
Omgivande |
UN:
1170 ADR: 3,III |