Pallets and sumps for drums and containers
Leverantör: asecos
2516 2520 2521 9843 9844 2500 2504 2508 2510 2512 2514 2501 2505 2509 2511 2513 2515
MSGA2516EA 0 SEK
MSGA2516 MSGA2520 MSGA2521 MSGA9843 MSGA9844 MSGA2500 MSGA2504 MSGA2508 MSGA2510 MSGA2512 MSGA2514 MSGA2501 MSGA2505 MSGA2509 MSGA2511 MSGA2513 MSGA2515
Pallets and sumps for drums and containers
Förvaring av tunnor
Available either painted (RAL 5010Gentian blue) or hot dip galvanised.
  • Direct storage in the sump pallet or on the grid
  • By adding drummounts, 60-litre and200-litre drums can be stored horizontally
  • Forklift truck pockets
  • Accessible from below on 4 sides
  • Easy and safe in-house transport
Approved for the safe storage of water pollutants and flammable liquids.