Wipes for general applications, WYPALL* X90
Leverantör: KIMBERLY CLARK
12891 12889
115-1532EA 1525 SEK
115-1532 115-1530
Wipes for general applications, WYPALL* X90
Torkdukar Torkdukar, återanvändbara Torkduk, universal
These soft yet durable wipes provide high grade oil and water absorption. They are ideal for heavy wiping, prepping surfaces with solvents, and cleaning metal shavings and rough surfaces.
  • More absorbent than other industrial wiping solutions and reusable: Less waste for users
  • Durable and strong thanks to two bonded plies of spunbond material
  • Available as a roll or in a portable dispenser
Colour: Blue