Syringe filter holders, Whatman™
Leverantör: Whatman products (Cytiva)
1980-001 1980-002
WHAT1980-001EA 2825 SEK
WHAT1980-001 WHAT1980-002
Syringe filter holders, Whatman™
Filterhållare
These syringe filter holders are made from stainless steel. The holders with Luer lock fittings connected to a standard syringe offer quick and easy clarification, sterilisation and removal of particulates from small volume samples, typically for HPLC applications.
They contain PTFE gaskets and O-rings which allow the membrane to be autoclaved in place without the filter sticking to the holder. Also available in PP.