Spill kits in overpack drum, PIG®
Leverantör: NEW PIG
KIT302 KITR302 KIT202-999 KIT402 RFL402
121-0509EA 15050 SEK
121-0509 121-0510 121-0155 121-0563 121-0158
Spill kits in overpack drum, PIG®
Spillkit
Spill kits containing all the absorbents needed to respond to a large spill of oils, coolants, solvents or water - all in one container that can then be used to dispose of the waste.
  • Leakproof, twist-on lid is notched so can easily be removed or tightened
  • Overpack is X-rated for Packing Groups I, II and III, so it's certified for shipping waste by land, sea or rail after cleanup is finished
  • Lift-out, prepacked baskets speed access and guard contents from UV
  • Ledges moulded into container make kit easy to move by forklift
  • Lightweight, 100% polyethylene container resists chemicals and keeps contents clean and dry
  • Bold 'Spill Kit' lettering for quick identification
  • Overpack protection cover (available separately) is suggested for outdoor storage to protect container from UV degradation and weathering